Skip to Content
Merck
All Photos(1)

Key Documents

MET58C

Supelco

2-Chloro-4,6-diamino-1,3,5-triazine

analytical standard

Synonym(s):

Atrazine-desethyl-desisopropyl, 6-Chloro-2,4-diamino-1,3,5-triazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H4ClN5
CAS Number:
Molecular Weight:
145.55
Beilstein:
124340
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

packaging

ampule of 100 mg

manufacturer/tradename

Chem Service, Inc. MET-13213C

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Nc1nc(N)nc(Cl)n1

InChI

1S/C3H4ClN5/c4-1-7-2(5)9-3(6)8-1/h(H4,5,6,7,8,9)

InChI key

FVFVNNKYKYZTJU-UHFFFAOYSA-N

Gene Information

mouse ... Esr1(13982)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joseane M Pozzebon et al.
Journal of chromatography. A, 987(1-2), 375-380 (2003-03-05)
A method was developed to determine simazine, atrazine and their metabolite, 2-chloro-4,6-diamino-1,3,5-triazine, in urine. The presence of these herbicides in urine may reflect possible exposure to pesticides. Sample preparation involved protein precipitation and solid-phase extraction. The samples were analyzed by
Jennifer L Seffernick et al.
Applied and environmental microbiology, 68(9), 4672-4675 (2002-08-30)
2-Chloro-4,6-diamino-s-triazine (CAAT) is a metabolite of atrazine biodegradation in soils. Atrazine chlorohydrolase (AtzA) catalyzes the dechlorination of atrazine but is unreactive with CAAT. In this study, melamine deaminase (TriA), which is 98% identical to AtzA, catalyzed deamination of CAAT to
Greg P Dooley et al.
Chemical research in toxicology, 19(5), 692-700 (2006-05-16)
Atrazine (2-chloro-4-[ethylamino]-6-[isopropylamino]-1,3,5-triazine) is one of the most commonly used herbicides in North America and is frequently detected in ground and surface waters. This research investigated possible covalent modifications of hemoglobin following in vivo exposures to atrazine in Sprague Dawley (SD)
C Ernst et al.
Applied microbiology and biotechnology, 42(5), 763-768 (1995-01-01)
A bacterium utilizing 2-chloro-4,6-diamino-s-triazine (CAAT) as sole nitrogen source was isolated under a N2-free atmosphere and identified as Klebsiella pneumoniae. Concomitant to CAAT degradation the protein content increased and chloride was released into the medium. Under air and a N2-atmosphere
Tami S McMullin et al.
Toxicology, 240(1-2), 1-14 (2007-09-05)
Atrazine (ATRA) is metabolized by cytochrome P450s to the chlorinated metabolites, 2-chloro-4-ethylamino-6-amino-1,3,5-triazine (ETHYL), 2-chloro-4-amino-6-isopropylamino-1, 3, 5-triazine (ISO), and diaminochlorotriazine (DACT). Here, we develop a set of physiologically based pharmacokinetic (PBPK) models that describe the influence of oral absorption and oxidative

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service