Skip to Content
Merck
All Photos(1)

Key Documents

LMPS140

Polysulfide in light mineral oil

0.5000% (5000 ppm), certified reference material (UKAS ISO 17025 & ISO 17034 certified)

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
R$1,232.00

R$1,232.00


Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
100 ML
R$1,232.00

About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

R$1,232.00


Estimated to ship onJune 01, 2025


Request a Bulk Order

grade

certified reference material (UKAS ISO 17025 & ISO 17034 certified)

Quality Level

form

liquid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

concentration

0.5000% (5000 ppm)

SMILES string

S(SC(C(C(C)(C)C)C)(C)C)SC(C(C(C)(C)C)C)(C)C

InChI

1S/C18H38S3/c1-13(15(3,4)5)17(9,10)19-21-20-18(11,12)14(2)16(6,7)8/h13-14H,1-12H3

InChI key

AVVJXRYTKMKDQS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service