Skip to Content
Merck
All Photos(1)

Key Documents

K08971330

Kromasil® 4 Column SFC Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

availability

available only in USA, Canada and Puerto Rico

technique(s)

HPLC: suitable

Footnote

4 Column Kit includes one each of the following columns.

K08971322 - Kromasil 2.5 micron Silica SFC HPLC Column - 150mm x 3.0mm
K08971324 - Kromasil 2.5 micron Diol SFC HPLC Column - 150mm x 3.0mm
K08971326 - Kromasil 2.5 micron CN SFC HPLC Column - 150mm x 3.0mm
K08971328 - Kromasil 2.5 micron 2EP SFC HPLC Column - 150mm x 3.0mm

Legal Information

Kromasil is a registered trademark of EKA Chemicals AB

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service