Skip to Content
Merck
All Photos(1)

Key Documents

G006165

LC/MS Analysis of Fluoroamphetamine Drugs of Abuse on Ascentis® Express HILIC

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

column: Ascentis Express HILIC, 10 cm x 2.1 mm I.D., 2.7 μm particles (53939-U)
gradient: 5 mM ammonium formate in acetonitrile:water (90:10, v/v)
flow rate: 0.4 mL/min
column temp.: 35 °C
detector: TOF/MS
injection: 2 μL
sample: 300 ng/mL in acetonitrile

suitability

application for HPLC

application(s)

forensics and toxicology

Analysis Note

The fluoroamphetamines (FA) are a group of "designer" stimulant drugs in amphetamine family. The rapid LC/MS analysis of four members of the FA family on Ascentis Express HILIC is shown here. Cerilliant CRMs were used to ensure reliable MS identification and quantification.

Legal Information

Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    (±)-2-Fluoroamphetamine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
    4-Fluoroamphetamine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
    (±)-2-Fluoromethamphetamine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
    (±)-4-Fluoromethamphetamine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service