Skip to Content
Merck
All Photos(1)

Documents

G005971

Stability and Reproducibility of Zenix® 300 under High Salt and High pH Mobile Phase Conditions

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

column: Zenix SEC-300, 30 cm x 7.8 mm I.D., 3 μm particles (Z777018)
mobile phase: 50 mM sodium phosphate + 1 M NaCl, pH 8.5
flow rate: 1.0 mL/min
pressure: 1450 psi (100 bar)
column temp.: 25 °C
detector: UV, 214 nm
injection: 5 μL
sample: each compound 1 mg/mL except uracil, 0.1 mg/mL

suitability

application for HPLC

application(s)

life science and biopharma

Legal Information

Zenix is a registered trademark of Sepax Technologies

Analyte

Description

    Thyroglobulin 1 mg/mL
    Bovine serum albumin (BSA) 1 mg/mL
    Ribonuclease A 1 mg/mL
    Uracil 1 mg/mL

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service