Skip to Content
Merck
All Photos(1)

Key Documents

G005889

HPLC Analysis of a Monoclonal Antibody on Zenix® SEC-150, 30 cm X 4.6 mm I.D., 3 μm particles after Proteolysis with IdeS Protease

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

column: Zenix® SEC-150, 30 cm X 4.6 mm I.D., 3 μm particles (Z777018)
mobile phase: 0.15 M potassium phosphate monobasic, pH 7.0 with potassium hydroxide
flow rate: 0.35 mL/min
pressure: 1450 psi (100 bar)
column temp.: ambient
detector: UV, 215 nm
injection: 1 μL
sample: digested SO57 mAb, ~0.5 g/L

suitability

application for HPLC

application(s)

life science and biopharma

Analysis Note

Proteolysis of antibodies can be useful for simplifying further analysis. This application demonstrates separation of the two fragments, F(ab′)2 and scFc, after digestion with IdeS protease

Legal Information

Zenix is a registered trademark of Sepax Technologies

related product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service