Skip to Content
Merck
All Photos(1)

Key Documents

G005601

HPLC Analysis of Levothyroxine and Liothyronine on Ascentis® Express ES-Cyano

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

Pricing and availability is not currently available.

technique(s)

HPLC: suitable

test parameters

column: Ascentis Express ES-Cyano, 10 cm x 2.1 mm I.D., 2.7 μm particles (53491-U)
mobile phase: [A] water with 0.05% phosphoric acid; [B] acetonitrile with 0.05% phophoric acid; (60:40, A:B)
flow rate: 1.5 mL/min
pressure: 3270 psi (225 bar)
column temp.: 30 °C
detector: UV, 225 nm
injection: 5 μL
sample: 20 μg/mL in mobile phase

suitability

application for HPLC

application(s)

clinical

Analysis Note

This application demonstrates the suitabilty of Ascentis Express ES-Cyano for the analysis of Levothyroxine according to USP Monograph 3301 with Liothyronine as a reference standard.

Legal Information

Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    3,3′,5-Triiodo-L-thyronine sodium salt, powder, BioReagent, suitable for cell culture
    Levothyroxine sodium hydrate, PESTANAL®, analytical standard

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service