Skip to Content
Merck
All Photos(1)

Key Documents

G004869

GC Analysis of ‘Positive List′ Chemicals (Mix 5) on SLB®-5ms

suitable for GC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

gas chromatography (GC): suitable

test parameters

column: SLB-5ms, 30 m x 0.25 mm I.D., 0.25 μm (28471-U)
oven: 80 °C (2 min), 5 °C/min to 300 °C (4 min)
inj. temp.: 250 °C
MSD interface: 300 °C
scan range: m/z 40-450
carrier gas: helium, 1 mL/min constant
injection: 1.0 μL splitless (1 min)
sample: each pesticide at 20 ppm in acetone

application(s)

agriculture
food and beverages

Legal Information

SLB is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    3,6-Bis(2-chlorophenyl)-1,2,4,5-tetrazine
    Biphenyl
    α-1,2,3,4,5,6-Hexachlorocyclohexane
    β-BHC
    Swep
    Lindane
    Pyroquilon
    δ-BHC
    MCPA-thioethyl
    Oxabetrinil
    Propanil
    Metribuzin
    Simetryn
    Ametryn
    Dichlofluanid
    Bromacil
    Tolylfluanid
    Captan
    Folpet
    Methoprene
    Chinomethionate
    Trichlamide
    Fludioxonil
    Nitrofen
    Oxadixyl
    Natriuretic Peptide, C-Type
    Piperonylbutoxide
    Nitralin
    [3-(3,5-Dichlorophenyl)-2,4-dioxoimidazolidinyl]-N-(methylethyl)carboxamide
    Bifenazate
    Pentoxazone
    Mefenacet
    Fenoxaprop-ethyl
    Bitertanol
    Silafluofen
    Pyrazoxyfen
    Famoxadone
    See All (37)

analytical column


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service