Skip to Content
Merck
All Photos(1)

Key Documents

CRM63613

Supelco

Nitrofen

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

2,4-Dichlorophenyl 4-nitrophenyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
R$905.00

R$905.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
R$905.00

About This Item

Empirical Formula (Hill Notation):
C12H7Cl2NO3
CAS Number:
Molecular Weight:
284.09
Beilstein:
1887356
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

R$905.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

format

neat

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1ccc(Oc2ccc(Cl)cc2Cl)cc1

InChI

1S/C12H7Cl2NO3/c13-8-1-6-12(11(14)7-8)18-10-4-2-9(3-5-10)15(16)17/h1-7H

InChI key

XITQUSLLOSKDTB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.comNitrofen is a diphenyl ether herbicide, used for the pre- and post-emergence control of annual grasses and broad-leafed weeds. It targets protoporphyrinogen oxidase (PPO) and causes the accumulation of photosensitizing porphyrins, which in turn lead to lipid peroxidation and membrane disruption.
Nitrofen is not approved for its use in the European Union (EU) as an active substance in plant protection products, according to Regulation (EC) No. 1107/2009. But a default maximum residue limit (MRL) of 0.01-0.02 mg/kg is applicable for its presence in products of plant and animal origin following the regulation (EC) No 149/2008.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
Nitrofen CRM may also be used as given below:

  • Development of a binary solvent-liquid phase microextraction (BS-LPME) method for sample treatment followed by gas chromatographic-mass spectrometric (GC-MS) analysis of alachlor, nitrofen, and bifenox in well, dam lake, and river water samples
  • Analysis of 10 samples of fruits and vegetables for the presence of 11 pesticide residues by combining TpPaNO2 (Tp stands for 1,3-triformylphloroglucinol and Pa-NO2 stands for 2-nitro-1,4-phenelynediamine) fiber-based solid-phase microextraction with gas chromatography-electron capture detection (GC-EID)
  • Evaluating the use of triazine-based conjugated microporous polymer (TCMP) as a fiber coating for the solid-phase microextraction (SPME) of 12 halogens-containing environmental hormones from three fruits and five vegetable samples for their determination by gas chromatography-electron capture detection (GC-ECD)
  • Multi-residue determination of 27 pesticides from different chemical classes in wine samples using dispersive liquid-liquid microextraction (DLLME) and gas chromatography-mass spectrometry (GC-MS)
  • Quantitative analysis of 113 pesticides and their metabolites in different fruit and vegetable samples using gas chromatography-tandem mass spectrometry (GC-MS/MS) with deactivated programmable temperature vapourising (PTV) injector
  • Simultaneous determination of 133 pesticides in commercially available chenpi samples by a combination of modified QuEChERS method and gas chromatography-tandem mass spectrometry (GC-MS/MS)

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service