Skip to Content
Merck
All Photos(1)

Documents

CRM44893

Supelco

ASTM® D6584 Monoolein solution

certified reference material, 5000 μg/mL in pyridine

Synonym(s):

Monoolein solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

Agency

ASTM® D6584

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 3 mL

concentration

5000 μg/mL in pyridine

technique(s)

gas chromatography (GC): suitable

application(s)

petroleum

format

single component solution

storage temp.

2-8°C

InChI

1S/C21H40O4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(24)25-19-20(23)18-22/h9-10,20,22-23H,2-8,11-19H2,1H3/b10-9-

InChI key

RZRNAYUHWVFMIP-KTKRTIGZSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany
ASTM is a registered trademark of American Society for Testing and Materials

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

62.6 °F - closed cup

Flash Point(C)

17.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service