Skip to Content
Merck
All Photos(1)

Key Documents

CRM40392

Supelco

Cinnamaldehyde solution

certified reference material, 2000 μg/mL in methanol, ampule of 1 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12164502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

product line

TraceCERT®

form

liquid

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

2000 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable

application(s)

cannabis testing

format

single component solution

storage temp.

-10 to -25°C

InChI

1S/C9H8O/c10-8-4-7-9-5-2-1-3-6-9/h1-8H/b7-4+

InChI key

KJPRLNWUNMBNBZ-QPJJXVBHSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alžbeta Líšková et al.
ALTEX, 37(3), 441-450 (2020-03-01)
Despite a continuous increase of commercial products containing nanoparticles, only few materials are currently used in such large amounts, forms and sizes as titanium dioxide - TiO2. Besides its use in cosmetics, food industry and biomedicine, TiO2 nanoparticles (NPs) are
I Radsetoulalova et al.
British poultry science, 61(2), 169-172 (2019-12-18)
1. The aim was to evaluate the acaricidal effects of pure active components of essential oils against poultry red mite (Dermanyssus gallinae) as an alternative to chemical acaricides (organophosphates and pyrethroids).2. The toxicities of five pure active components of essential
Angel Llopis-Giménez et al.
Journal of chemical ecology, 46(11-12), 1017-1031 (2020-11-06)
Insect chemosensation is crucial for many aspects related to food seeking, enemy avoidance, and reproduction. Different families of receptors and binding proteins interact with chemical stimuli, including odorant receptors (ORs), ionotropic receptors (IRs), gustatory receptors (GRs), odorant binding proteins (OBPs)
Fenxia Hou et al.
ALTEX, 37(3), 451-468 (2020-04-24)
There are currently no skin sensitization assays based on T cell activation. We built a novel in vitro model to assess T cell activation and test its performance to discriminate skin sensitizers from non-sensitizers using 52 reference chemicals. Jurkat Clone
C A Mini et al.
Toxicology in vitro : an international journal published in association with BIBRA, 71, 105069-105069 (2020-12-15)
Three-dimensional skin models, also named 3D skin models, human skin equivalents (HSEs), or Human Epidermal Equivalents (HEEs), have been increasingly used for chemical assessments in terms of efficacy and safety. Considering this, we developed an HEE model using immortalized HaCaT

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service