91225
1,3-Butanediol dimethacrylate
analytical standard
About This Item
Recommended Products
grade
analytical standard
Assay
≥94.0% (GC)
form
liquid
shelf life
limited shelf life, expiry date on the label
contains
100-300 ppm MEHQ as inhibitor
refractive index
n20/D 1.450-1.454
n20/D 1.452 (lit.)
bp
290 °C (lit.)
density
1.01 g/mL at 25 °C (lit.)
SMILES string
CC(CCOC(=O)C(C)=C)OC(=O)C(C)=C
InChI
1S/C12H18O4/c1-8(2)11(13)15-7-6-10(5)16-12(14)9(3)4/h10H,1,3,6-7H2,2,4-5H3
InChI key
VDYWHVQKENANGY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Recommended products
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service