Skip to Content
Merck
All Photos(1)

Key Documents

821816

TSKgel® SuperMPHZ-N (3 µm) HPLC Columns

L x I.D. 2 cm × 4.6 mm, polystyrene-divinylbenzene phase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711

Product Name

TSKgel® SuperMPHZ-N Guard Column, phase polystyrene-divinylbenzene, L × I.D. 2 cm × 4.6 mm, 3 μm particle size

material

stainless steel

product line

TSKgel®

manufacturer/tradename

Tosoh 21816

parameter

0.4 mL/min flow rate
60 °C temperature

technique(s)

HPLC: suitable

L × I.D.

2 cm × 4.6 mm

matrix

polystyrene-divinylbenzene

matrix active group

polystyrene-divinylbenzene phase

particle size

3 μm

separation technique

size exclusion (SEC)

General description

Tosoh′s innovative multi-pore technology is now also available in 3, 4 and 6 micron TSKgel® SuperMultiporeHZ columns and guard columns. Three columns, varying in linear range, are available. The base material of each TSKgel SuperMultiporeHZ column is poly(styrene/divinylbenzene). Particles in TSKgel SuperMultiporeHZ columns and guard columns are mono-disperse in size and exhibit a broad range of pore sizes. Each particle, by design, has an extended linear calibration curve, thereby greatly diminishing the presence of inflection points in chromatograms.

Physical form

Column can only be used in THF solvent.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service