821463
TSKgel® BioAssist® Desalting (15 µm) HPLC Columns
L x I.D. 15 cm × 4.6 mm, polyacrylamide phase
About This Item
Recommended Products
product name
TSKgel® BioAssist® Desalting HPLC Column, phase polyacrylamide, L × I.D. 15 cm × 4.6 mm, 15 μm particle size, PEEK
material
PEEK
product line
TSKgel®
manufacturer/tradename
Tosoh 21463
parameter
≤30% polar organic solvent
1.5 mL/min flow rate
13 bar max. pressure
4-30 °C temperature
technique(s)
HPLC: suitable
L × I.D.
15 cm × 4.6 mm
matrix
urea cross-linked polyacrylamide
matrix active group
polyacrylamide phase
particle size
15 μm
operating pH
3.0-12.0(<= 1 wk)
4.0-11.0(> 1 wk)
separation technique
size exclusion (SEC)
Looking for similar products? Visit Product Comparison Guide
General description
The novel, hydrophilic, highly cross-linked polyacrylamide beads exhibit superior mechanical strength compared with conventional hydrophilic polyacrylamide beads and cross-linked dextran beads. This increase in strength is what allows the use of the small spherical 15μm beads. TSKgel BioAssist DS columns can be operated in standard HPLC systems to quickly and efficiently reduce salt and/or buffer concentrations of collected protein or nucleic acid fractions.
Physical form
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service