Skip to Content
Merck
All Photos(1)

Key Documents

821411

TSKgel® BioAssist® S (13 µm) HPLC Columns

L x I.D. 10 cm × 10 mm, sulfonic acid phase

Synonym(s):

TSKgel® BioAssist® S HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
eCl@ss:
32110501
Pricing and availability is not currently available.

Product Name

TSKgel® BioAssist® S HPLC Column, phase sulfonic acid, L × I.D. 10 cm × 10 mm, 13 μm particle size, PEEK hardware

material

PEEK hardware

product line

TSKgel®

feature

counter ion Na+

manufacturer/tradename

Tosoh 21411

parameter

≤2.6 M salt concentration
≤30% organic solvent
25 bar max. pressure
4-60 °C temperature
5 mL/min flow rate

technique(s)

HPLC: suitable

L × I.D.

10 cm × 10 mm

matrix

polymer particle platform

matrix active group

sulfonic acid phase

particle size

13 μm

pore size

1300 Å

operating pH

2-12(<= 1 month)
3-10(> 1 month)

separation technique

cation exchange

Looking for similar products? Visit Product Comparison Guide

General description

TSKgel BioAssist S columns are packed with spherical, polymer beads with approx. 1300Ã… pores. They are surface modified with a strong sulfopropyl cation exchange groups. The large pores allow large biomolecules to freely enter and exit the pores resulting in higher binding capacity and better recovery of activity than other competivie materials. Standard column dimensions are 4.6mm ID x 5cm, while a 10mm ID x 10cm semi-preparative column is available for scale-up. Both columns are made of PEEK to reduce protein adsorption.

Physical form

Shipped in 20% Ethanol in 20 mM phosphate buffer (pH 6.5).

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

BioAssist is a registered trademark of Tosoh Corporation
TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service