821391
ToyoScreen® AF-Heparin HC-650M Process Development Column
phase sulfated glycosaminoglycan, L × I.D. 3 cm × 14.6 mm, 65 μm particle size
About This Item
material
aluminum seal
polypropylene and polyethylene frit
manufacturer/tradename
Tosoh 21391
parameter
1-5 mL/min flow rate
20 mL/min max. flow rate
3 bar max. pressure
L × I.D.
3 cm × 14.6 mm
matrix
hydroxylated methacrylic polymer
matrix active group
sulfated glycosaminoglycan phase
particle size
65 μm
operating pH
5-10
capacity
>5 g/L adsorption capacity (antithrombin III)
separation technique
affinity
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service