Skip to Content
Merck
All Photos(1)

Key Documents

819308

Holder for 2 mm I.D. TSKgel® HPLC Guard Cartridges

pkg of 1 ea

Synonym(s):

TSKgel® Holder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115716
eCl@ss:
32110501
Pricing and availability is not currently available.

Product Name

Holder for 2 mm I.D. TSKgel® Guard Cartridges, pkg of 1 ea

material

stainless steel

product line

TSKgel®

packaging

pkg of 1 ea

manufacturer/tradename

Tosoh 19308

technique(s)

HPLC: suitable

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service