Skip to Content
Merck
All Photos(1)

Key Documents

813727

TSKgel® HXL (13 µm) HPLC Columns

L x I.D. 4 cm × 6 mm, polystyrene-divinylbenzene phase

Synonym(s):

TSKgel® G1000HXL HPLC Column for Light Scattering Applications

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
Pricing and availability is not currently available.

Product Name

TSKgel® HXL Guard Column, phase polystyrene-divinylbenzene, L × I.D. 4 cm × 6 mm, 13 μm particle size

material

stainless steel

product line

TSKgel®

manufacturer/tradename

Tosoh 13727

parameter

1.2 mL/min flow rate
60 °C temperature

technique(s)

HPLC: suitable

L × I.D.

4 cm × 6 mm

matrix

polymer particle platform

matrix active group

polystyrene-divinylbenzene phase

particle size

13 μm

pore size

mixed

separation technique

size exclusion (SEC)

General description

Guard column to protect G5000HXL - G6000HXL columns.

Physical form

Shipped in THF.

Other Notes

THF can be replaced with benzene, chloroform, toluene, xylene, dichloromethane, dichloroethane

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service