Skip to Content
Merck
All Photos(2)

Key Documents

67709

Supelco

N-Nitroso-N,N-bis(7-methyloctyl)amine

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H38N2O
CAS Number:
Molecular Weight:
298.51
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-8°C

SMILES string

O=NN(CCCCCCC(C)C)CCCCCCC(C)C

InChI

1S/C18H38N2O/c1-17(2)13-9-5-7-11-15-20(19-21)16-12-8-6-10-14-18(3)4/h17-18H,5-16H2,1-4H3

InChI key

XAYFTZOFOLGWAE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service