Skip to Content
Merck
All Photos(1)

Key Documents

59465-U

Opti-Max® inlet check valve

for use with LC-6A, LC-10AS, Shimadzu 228-12353-91

Synonym(s):

Optimize Technologies HPLC Pump Part for Shimadzu

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854
NACRES:
SB.51

form

solid

packaging

pkg of 1

manufacturer/tradename

Shimadzu 228-12353-91

compatibility

for use with LC-6A, LC-10AS

Looking for similar products? Visit Product Comparison Guide

Legal Information

Opti-Max is a registered trademark of Optimize Technologies, Inc.
Shimadzu is a trademark of Shimadzu Corporation

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service