Skip to Content
Merck
All Photos(1)

Key Documents

58997C30

Supelco

SUPELCOSIL LC-SCX (5 µm) HPLC Columns

L × I.D. 25 cm × 3 mm, HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

Product Name

SUPELCOSIL LC-SCX HPLC Column, 5 μm particle size, L × I.D. 25 cm × 3 mm

material

stainless steel column

Agency

suitable for USP L52

product line

SUPELCOSIL

feature

endcapped: no

manufacturer/tradename

SUPELCOSIL

packaging

1 ea of

parameter

≤70 °C temp. limit
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

25 cm × 3 mm

surface area

170 m2/g

matrix

silica gel, spherical particle platform
fully porous particle

matrix active group

sulfonic acid, propyl- phase

particle size

5 μm

pore size

120 Å

pH range

2-7.5

application(s)

food and beverages

separation technique

cation exchange

Looking for similar products? Visit Product Comparison Guide

General description

The LC-SCX cation-exchange columns have strongly acidic propylsulfonic acid groups and are used for separating cations. Adjust pH, ionic strength, and organic modifier concentration to control retention and selectivity.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jie Zhao et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 33(15), 1853-1855 (2008-11-15)
To developed a method for the determination of thiabendazole residues in barbary wolfberry fruit by ion exchange chromatography (IEC) with fluorescence detection. The samples were extracted with 0.1% H3PO4. Chromatographic analysis was performed on a Supelcosil LC-SCX (4.6 mm x

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service