Skip to Content
Merck
All Photos(1)

Documents

58255

Column Frits

diam. 1/2 in., porosity 2 μm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

material

stainless steel frit

description

for Conventional Column Hardware (compression style)

packaging

pkg of 10 ea

diam.

1/2 in.

thickness

.043 in.

pore size

2 μm porosity

compatibility

for use with compression style column hardware

Looking for similar products? Visit Product Comparison Guide

General description

Choose frits with 0.5 μm pores to retain packings prepared from 3 μm particles, frits with 2 μm pores to retain larger particles.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service