Skip to Content
Merck
All Photos(1)

Key Documents

57648

Sample Loop for 9010, 9125, and 9725 Injectors

volume 2 mL, PEEK

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854
Pricing and availability is not currently available.

material

PEEK
PEEK sample loop

O.D.

1.6 mm
1/16 in.

volume

2 mL

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service