Key Documents
57625
Sample Loop for Model 7725 Injector
volume 20 μL
About This Item
UNSPSC Code:
41115854
Pricing and availability is not currently available.
volume
20 μL
General description
Sample loop for model 7725 injector (volume 20 μL) is designed for advanced fluid-handling solutions for a variety of analytical instrumentation and in vitro diagnostic systems. It can be employed in high performance liquid chromatographic (HPLC) systems and is quite accurate and precise in injecting 1 μL-5 mL concentration of samples.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Articles
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.