Key Documents
57476
Needle for Model 3725i Prep Sample Injector
stainless steel, needle size 16 ga
About This Item
UNSPSC Code:
41115854
material
stainless steel
needle size
16 ga
packaging
pkg of 1
compatibility
for use with Rheodyne 3725-086
General description
Needle for Model 3725i Prep Sample Injector includes a 5 cm large-bore syringe needle (16 gauge/0.65 in. OD) for rapid injection of large samples.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Choose from one of the most recent versions:
Certificates of Analysis (COA)