Skip to Content
Merck
All Photos(1)

Documents

54775

Supelco

LiChrospher® RP-18 (5 µm) HPLC Columns

L × I.D. 15 cm × 3.2 mm, HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

product name

LiChrospher® RP-18 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 3.2 mm

Agency

suitable for USP L1

manufacturer/tradename

LiChrospher®

extent of labeling

21% Carbon loading

parameter

≤70 °C temp. range
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

15 cm × 3.2 mm

surface area

350 m2/g

matrix

octadecyl
silica particle platform

matrix active group

C18 (octadecyl) phase

particle size

5 μm

pore size

100 Å

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

LiChrospher® is a reliable and versatile spherical silica sorbent, which is traditionally produced and available with different modifications. LiChrospher® RP-18 (5 μm particle size, L × I.D. 15 cm × 3.2 mm) delivers highly reproducible results and is designed for reversed-phase chromatography of acidic, neutral and weakly basic compounds.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChrospher is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service