Skip to Content
Merck
All Photos(1)

Key Documents

54281-U

Supelco

Supelclean Ultra 2400 solid phase extraction (SPE) Tubes

volume 3 mL, pkg of 54

Synonym(s):

Supelclean Ultra

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115712
NACRES:
NB.21

product name

Supelclean Ultra 2400, volume 3 mL, pkg of 54

product line

Supelclean

Quality Level

composition

bed A, 270 mg
bed B, 225 mg (Z-Sep)

packaging

pkg of 54

technique(s)

solid phase extraction (SPE): suitable

volume

3 mL

matrix active group

C18 bonding
PSA bonding
carbon bonding

application(s)

food and beverages

Looking for similar products? Visit Product Comparison Guide

General description

Supelclean PSA, Discovery® C18, and Spherical Carbon
This multi-bed solid phase extraction (SPE) cartridge was designed for the cleanup of extracts of difficult matrices such as dry commodities (tea, spices, coffee, etc.) prior to pesticide residue analysis, typically performed by GC/MSMS and LC/MS/MS. These types of samples can contain pigments and oils, and may not be sufficiently cleaned using a standard QuEChERS cleanup.
The top bed consists of a mixture of PSA/C18 and a graphitized, spherical carbon. PSA removes acidic interferences, C18 retains some hydrophobic interferences, and carbon removes pigments. The variety of carbon used in this cartridge was engineered to remove sufficient pigmentation while allowing for better recoveries of compounds with planar structures. The bottom bed of the cartridge contains Z-Sep, which is a zirconia-coated silica. Z-Sep removes oily residues, and provides additional pigment removal.
The cartridge can be used for cleanup of acetonitrile extracts to be analyzed by either GC or HPLC.

Legal Information

Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany
Supelclean is a trademark of Sigma-Aldrich Co. LLC

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Dual-layer cartridge achieves efficient SPE clean-up for challenging pesticide residue testing in turmeric and spices.

2-layer SPE cartridge efficiently reduces background for ginger contaminant analysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service