54036-U
pk20 Envicarb/LC-NH2/Si SPE Tubes 20mL
bed A 500 mg (Carbopack B 120/400), bed C 600 mg (Supelclean LC-Si), bed B 400 mg (Supelclean LC-NH2), volume 20 mL, pkg of 20 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41115712
NACRES:
NB.21
Recommended Products
material
polypropylene tube
composition
bed A, 500 mg (Carbopack B 120/400)
bed B, 400 mg (Supelclean LC-NH2)
bed C, 600 mg (Supelclean LC-Si)
packaging
pkg of 20 ea
technique(s)
solid phase extraction (SPE): suitable
volume
20 mL
matrix active group
amino phase
carbon phase
silica phase
application(s)
food and beverages
separation technique
normal phase
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service