52753-U
Discovery® DSC-Diol SPE Tube
bed wt. 1 g, volume 6 mL, pk of 30
About This Item
Recommended Products
material
polypropylene tube
Quality Level
composition
bed wt., 1 g
packaging
pk of 30
extent of labeling
7% C loading
technique(s)
solid phase extraction (SPE): suitable
surface area
480 m2/g
volume
6 mL
matrix
Silica gel base material (irregularly shaped, acid washed)
matrix active group
diol bonding, polymerically bonded
particle size
50 μm
pore size
0.9 mL/g pore volume
70 Å pore diameter
application(s)
food and beverages
separation technique
normal phase
Looking for similar products? Visit Product Comparison Guide
General description
Sample Matrix Compatibility: Organic solvents, oils, and lipids
- Polymerically bonded, 2,3-Dihydroxypropoxypropyl (7% C)
- Polar sorbent most commonly used for normal phase applications (polar extractions from non-polar matrices)
- The sorbent dihydroxy groups facilitate strong hydrogen bonding
- Excellent selectivity when extracting structurally similar molecules
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
STOT SE 2 Inhalation
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service