Skip to Content
Merck
All Photos(1)

Key Documents

506761

Supelco

4-tert-Butylphenol

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CC6H4OH
CAS Number:
Molecular Weight:
150.22
Beilstein:
1817334
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

vapor pressure

1 mmHg ( 70 °C)

product line

PESTANAL®

CofA

current certificate can be downloaded

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

236-238 °C (lit.)

mp

96-101 °C (lit.)

density

0.908 g/mL at 25 °C (lit.)

application(s)

agriculture
cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-30°C

SMILES string

CC(C)(C)c1ccc(O)cc1

InChI

1S/C10H14O/c1-10(2,3)8-4-6-9(11)7-5-8/h4-7,11H,1-3H3

InChI key

QHPQWRBYOIRBIT-UHFFFAOYSA-N

Gene Information

mouse ... Esr1(13982)
rat ... Ar(24208)

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 1 - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Sonnich Meier et al.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP, 145(3), 420-430 (2007-03-09)
Offshore oil production releases large amounts of lipophilic compounds in produced water into the ocean. In 2004, 143 million m(3) produced water, containing approximately 13 tons of long-chain (>C(4)) alkylphenols (AP), was discharged from installations in the Norwegian sector of
Lei Lei et al.
Ecotoxicology (London, England), 19(7), 1268-1276 (2010-06-17)
Landfill leachates contain a large amount of unknown harmful compounds derived from domestic and industrial sources. A toxicity effect-directed approach was used to identify biologically active compounds in three landfill leachate samples (S1-S3) by combining the Microtox test with reverse-phase
Pierre Etienne Chabrier et al.
CNS drug reviews, 13(3), 317-332 (2007-09-27)
BN82451 belongs to a new family of small molecules designated as multitargeting or hybrid molecules. BN82451 is orally active, has good central nervous system penetration, and elicits potent neuronal protection and antiinflammatory properties. Neuronal protection is due to Na+ channel
J Pablo Lamas et al.
Analytical and bioanalytical chemistry, 394(8), 2231-2239 (2009-07-07)
A method for the determination of dimethyl fumarate (DMF), benzothiazole (BT) and tert-butylphenol (TBP) in desiccant and antimould agents employed for protecting consumer products from humidity and mould has been developed. The method is based on ultrasound-assisted extraction (UAE) followed
Zhigang Xu et al.
Journal of chromatography. A, 1358, 52-59 (2014-07-20)
In this study, a novel dual-template molecularly imprinted polymer (MIP)-coated stir bar was prepared and coupled with high-performance liquid chromatography (HPLC) for the analysis of environmental estrogens in complex samples. Dual-template MIP coating was homogeneous and porous with an average

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service