Skip to Content
Merck
All Photos(1)

Key Documents

48874

Supelco

2-Bromoethanol solution

certified reference material, 2000 μg/mL in toluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2CH2OH
CAS Number:
Molecular Weight:
124.96
UNSPSC Code:
12000000

grade

certified reference material
TraceCERT®

Agency

ASTM® D5578

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

2000 μg/mL in toluene

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

InChI

1S/C2H5BrO/c3-1-2-4/h4H,1-2H2

InChI key

LDLCZOVUSADOIV-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany
ASTM is a registered trademark of American Society for Testing and Materials

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.2 °F - closed cup

Flash Point(C)

4.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Min et al.
Annales pharmaceutiques francaises, 47(5), 266-275 (1989-01-01)
The mutagenic potencies of 13 bromoethyl esters of natural organic acids, have been studied, by Ames's test (strains TA 98 and TA 100, with and without system of metabolisation, S9 mix). None of the 8 bromoethyl esters of linoleic, oleic
Caroline C Womack et al.
The journal of physical chemistry. A, 115(51), 14559-14569 (2011-11-11)
This work characterizes the internal energy distribution of the CD(2)CD(2)OH radical formed via photodissociation of 2-bromoethanol-d(4). The CD(2)CD(2)OH radical is the first radical adduct in the addition of the hydroxyl radical to C(2)D(4) and the product branching of the OH
S Ebina et al.
Experientia, 36(5), 534-535 (1980-05-15)
2-Halogeno-ethanols change the active site structure of alpha-chymotrypsin more rapidly and effectively than ethanol, 1-propanol and urea, probably before producing an extensive conformation change.
S Khan et al.
Biochemical pharmacology, 45(2), 439-447 (1993-01-26)
The cytotoxicity of dibromoalkanes to isolated hepatocytes was proportional to the dibromoalkane concentration and increasing chain length of the dibromoalkane (C2-C6). The rapid hepatocyte glutathione (GSH) depletion which occurred upon addition of the dibromoalkanes was also dependent on the concentration
H Dunkelberg
Zentralblatt fur Bakteriologie, Mikrobiologie und Hygiene. 1. Abt. Originale B, Hygiene, 177(3-4), 269-281 (1983-04-01)
The compounds 2-chloroethanol and 2-bromoethanol were administered once weekly subcutaneously to groups of 100 female NMRI mice, in the case of 2-chloroethanol in 3 dosages (3.0, 1.0 and 0.3 mg, single dosage per mouse) and 2-bromoethanol in 2 dosages (1.0

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service