Skip to Content
Merck
All Photos(1)

Key Documents

48517

Supelco

2-Chloronaphthalene

for spectrophotometric det. of proline and thiophene, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7Cl
CAS Number:
Molecular Weight:
162.62
EC Number:
UNSPSC Code:
12000000

Assay

≥99.0%

CofA

current certificate can be downloaded

quality

for spectrophotometric det. of proline and thiophene

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

storage temp.

2-30°C

InChI

1S/C10H7Cl/c11-10-6-5-8-3-1-2-4-9(8)7-10/h1-7H

InChI key

CGYGETOMCSJHJU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sun-Hee Hyun et al.
PloS one, 8(9), e73065-e73065 (2013-09-24)
The metabolic profiles of Cordyceps bassiana according to fruiting body developmental stage were investigated using gas chromatography-mass spectrometry. We were able to detect 62 metabolites, including 48 metabolites from 70% methanol extracts and 14 metabolites from 100% n-hexane extracts. These
Barış Güzel et al.
Journal of mass spectrometry : JMS, 55(12), e4672-e4672 (2020-11-21)
Nowadays, a wide variety of chemicals has emerged because of increasing industrial and human activities. Thus, environmental chemistry laboratories performing analytical measurements need rapid and sensitive methods more than ever to get reliable quantitative data. For this purpose, a purge
Junsang Oh et al.
Journal of microbiology (Seoul, Korea), 57(1), 54-63 (2018-12-31)
Cordyceps militaris is a species of Cordyceps that is classified in the Cordycipitaceae family and is well known in East Asia as a traditional medicinal mushroom. Its artificial fruit body has been widely cultivated for commercial use in cosmetics, functional
Alok Sinha et al.
Journal of colloid and interface science, 314(2), 552-561 (2007-06-19)
Interaction of 2-chloronaphthalene (2-CN) with high-carbon iron filings (HCIF) was studied in anaerobic batch systems, both under well-mixed and poorly-mixed conditions. In well-mixed conditions, partitioning of 2-CN between solid and aqueous phases was fast, resulting in rapid attainment of equilibrium.
Chao Wang et al.
World journal of microbiology & biotechnology, 33(8), 160-160 (2017-07-29)
The lipid productivity controlled by both of biomass and lipid content was really crucial for economic-feasibility of microalgae-based biofuels production. This study attempted at augmenting lipid productivity in an emerging oleaginous model alga Coccomyxa subellipsoidea by different nitrogen manipulation including

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service