Skip to Content
Merck
All Photos(1)

Key Documents

47913

Supelco

EPA 8080 Pesticides Mix

certified reference material, each component in hexane: toluene (1:1) (varied)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

grade

certified reference material
TraceCERT®

Agency

EPA 8081

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

each component in hexane: toluene (1:1) (varied)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    α-BHC 250 μg/mL
    β-BHC 250 μg/mL
    Lindane 250 μg/mL
    δ-BHC 250 μg/mL
    Aldrin 250 μg/mL
    6-Hydroxy-2-naphthyl disulfide 250 μg/mL
    1,1-Dichloro-2,2-bis(4-chlorophenyl)ethene 250 μg/mL
    4,4′-DDT 250 μg/mL
    Dieldrin 250 μg/mL
    α-Endosulfan 250 μg/mL
    β-Endosulfan 250 μg/mL
    Endosulfan sulfate 250 μg/mL
    Endrin 250 μg/mL
    Endrin aldehyde 250 μg/mL
    Heptachlor 250 μg/mL
    Heptachlor exo-epoxide 250 μg/mL
    Methoxychlor 1000 μg/mL
    See All (17)

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-0.4 °F - closed cup

Flash Point(C)

-18 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service