Skip to Content
Merck
All Photos(1)

Key Documents

47867

Supelco

D-Pantothenic acid hemicalcium salt

analytical standard

Synonym(s):

(R)-(+)-N-(2,4-Dihydroxy-3,3-dimethyl-1-oxobutyl)-β-alanine hemicalcium salt, Calcium D-pantothenate, Vitamin B5

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2C(CH3)2CH(OH)CONHCH2CH2CO2 ·1/2Ca
CAS Number:
Molecular Weight:
238.27
Beilstein:
3769272
EC Number:
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

feature

standard type water soluble vitamin

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)
vitamins, nutraceuticals, and natural products

format

neat

storage temp.

2-8°C

SMILES string

CC(C)(CO)[C@@H](O)C(=O)NCCC(=O)O[Ca]OC(=O)CCNC(=O)[C@H](O)C(C)(C)CO

InChI

1S/2C9H17NO5.Ca/c2*1-9(2,5-11)7(14)8(15)10-4-3-6(12)13;/h2*7,11,14H,3-5H2,1-2H3,(H,10,15)(H,12,13);/q;;+2/p-2/t2*7-;/m00./s1

InChI key

FAPWYRCQGJNNSJ-UBKPKTQASA-L

Looking for similar products? Visit Product Comparison Guide

General description

D-Pantothenic acid hemicalcium salt is a vitamin present in several foods as free form and also bound in coenzyme A and acyl carrier protein.

Application

D-Pantothenic acid hemicalcium salt has been used as a calibration standard in the determination of vitamin B content present in samples of pineapple, using ultraviolet-visible light spectrophotometry.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Precursor in the biosynthesis of coenzyme A.

Physical properties

Due to the unstable, hygroscopic nature of the free acid, the calcium salt is employed.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of acoustically assisted treatments on vitamins, antioxidant activity, organic acids and drying kinetics of pineapple
Rodriguez O, et al.
Ultrasonics Sonochemistry, 35, 92-102 (2017)
Nicholas J Loman et al.
Nature reviews. Microbiology, 13(12), 787-794 (2015-11-10)
Twenty years ago, the publication of the first bacterial genome sequence, from Haemophilus influenzae, shook the world of bacteriology. In this Timeline, we review the first two decades of bacterial genome sequencing, which have been marked by three revolutions: whole-genome
Maha R Farhat et al.
Genome medicine, 6(11), 101-101 (2014-12-09)
Whole genome sequencing is increasingly used to study phenotypic variation among infectious pathogens and to evaluate their relative transmissibility, virulence, and immunogenicity. To date, relatively little has been published on how and how many pathogen strains should be selected for
Daniel R G Price et al.
BMC biology, 12, 110-110 (2014-12-21)
Genome evolution in intracellular microbial symbionts is characterized by gene loss, generating some of the smallest and most gene-poor genomes known. As a result of gene loss these genomes commonly contain metabolic pathways that are fragmented relative to their free-living
Henning Frerigmann et al.
Plant physiology, 166(1), 349-369 (2014-07-23)
By means of yeast (Saccharomyces cerevisiae) two-hybrid screening, we identified basic helix-loop-helix transcription factor05 (bHLH05) as an interacting partner of MYB51, the key regulator of indolic glucosinolates (GSLs) in Arabidopsis (Arabidopsis thaliana). Furthermore, we show that bHLH04, bHLH05, and bHLH06/MYC2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service