Skip to Content
Merck
All Photos(1)

Key Documents

46961

Supelco

Canola oil

analytical standard

Synonym(s):

Rapeseed oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12164500

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

General description

Characterized canola oil samples are available for use as controls or check samples for fatty acid methyl ester (FAME) analyses. These reference materials can be used as qualitative standards. They provide an excellent means of standardizing the lipid procedures and comparing the results to others. Packed in an amber ampule under nitrogen. A Certificate of Composition, which includes the chromatographic fingerprint analysis, is provided with the oil sample.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laziz Bouzidi et al.
Food & function, 4(1), 130-143 (2012-10-17)
Pure saturated triacylglycerols (TAGs) in canola oil were used as model systems to analyse oil loss in structured oil both from thermodynamic and kinetic perspectives. Two important parameters which effectively and predictively measure the relative propensity of a solid network
Ask the doctor. I use a lot of canola oil in cooking, because it's heart-healthy. My husband has prostate cancer, and I just heard that canola oil might make it worse. Is that really true and should I stop using it for cooking?
Anthony L Komaroff
Harvard health letter, 37(11), 2-2 (2012-10-19)
Cengizhan Yigitler et al.
Clinics (Sao Paulo, Brazil), 67(11), 1303-1308 (2012-11-28)
Postsurgical abdominal adhesions are common, serious postoperative complications. The present study compared the usefulness of 4% icodextrin and canola oil in preventing postoperative peritoneal adhesions. Twenty-four Wistar albino rats were divided into three groups. Following a laparotomy, a serosal abrasion
Jiang Jiang et al.
Meat science, 93(3), 469-476 (2013-01-01)
Alkali (pH(12)) and acid (pH(1.5)) pH-treated soy protein isolates (SPI) were incorporated (0.25-0.75% protein) into sols of myofibrillar protein (MP, 3%, in 0.6 M NaCl at pH 6.25) with or without 0.1% microbial transglutaminase (TG) to investigate the potential as
Nagao Totani et al.
Journal of oleo science, 61(11), 601-607 (2012-11-10)
We reported previously that in oils used for frying by commercial establishments, a high correlation was observed among their Gardner colors, polar compound contents (PC), carbonyl values (CV) and acid values (AV). However, this was not true for frying oils

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service