Skip to Content
Merck
All Photos(1)

Key Documents

46949

Supelco

Coconut oil

new

analytical standard

Synonym(s):

Coconut oil from Cocos nucifera, Coconut fat, Copra oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12164500

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

23-27 °C

application(s)

food and beverages

format

neat

storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

General description

Characterized coconut oil samples are available for use as controls or check samples for fatty acid methyl ester (FAME) analyses. These reference materials can be used as qualitative standards. They provide an excellent means of standardizing the lipid procedures and comparing the results to others. Packed in an amber ampule under nitrogen. A Certificate of Composition, which includes the chromatographic fingerprint analysis, is provided with the oil sample.
This reference material is offered as a qualitative standard. Use the chromatographic fingerprint of our reference material to check for the presence of coconut oil in your sample. A Certificate of Composition which includes a chromatogram is shipped with each product.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Does coconut oil reduce the risk of coronary artery diseases?
Ailya Inayat et al.
JPMA. The Journal of the Pakistan Medical Association, 63(6), 797-797 (2013-08-02)
Ask the doctor. Would you please comment on the current craze for coconut oil? I now have friends who, based on ads and popular books, are taking coconut oil several times a day.
Anthony L Komaroff
Harvard health letter, 37(12), 2-2 (2012-12-19)
Graciela E Hurtado de Catalfo et al.
Nutrition (Burbank, Los Angeles County, Calif.), 29(2), 462-469 (2012-12-04)
To investigate the effect of different dietary oils on the main hepatic enzymes involved in metabolism and their impact on oxidative stress status. Twenty-four male Wistar rats were fed for 60 d on the same basal diet plus different lipid
Jin-Sil Park et al.
Journal of medicinal food, 22(7), 672-679 (2019-05-22)
Dietary products may protect against inflammatory bowel disease (IBD) through mechanisms such as forming gut microbiota structures and providing substrates for microbial metabolism. Recently, many studies have been conducted on diets that potentially alleviate or suppress IBD development. To assess
Seyed Mohammad Taghi Gharibzahedi et al.
International journal of biological macromolecules, 51(4), 618-626 (2012-07-04)
An acceptable strategy to incorporate canthaxanthin (CX) as a natural colorant into products is by means of oil-in-water emulsions. The used CX in this study was produced by bacterium Dietzia natronolimnaea HS-1 using a batch bioreactor system. A central composite

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service