Skip to Content
Merck
All Photos(1)

Documents

44575-U

Supelco

Aliphatic Hydrocarbons Kit

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

CofA

current certificate can be downloaded

concentration

(34 individual ampules of neats plus 6 ampules of solutions)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

petroleum

format

multi-component solution

storage temp.

2-30°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    Decane 1 g
    1-Decene .5 g
    Docosane .1 g
    Dodecane 1 g
    1-Dodecene .5 g
    Dotriacontane .1 g
    1-Eicosene .5 g
    Heneicosane .1 g
    Heptane 1 g
    Hexacosane .1 g
    Hexadecane .1 g
    1-Hexadecene .5 g
    Hexane 1 g
    Hexatriacontane .1 g
    Nonane 1 g
    Octacosane 1 g
    Octadecane 1 g
    1-Octadecene .5 g
    Octane 1 g
    1-Octene .5 g
    Pentacosane .1 g
    Pentane 1 g
    Squalane 1 g
    Squalene 1 g
    Tetracontane .1 g
    Tetracosane 1 g
    Tetradecane 1 g
    1-Tetradecene .5 g
    Tetratetracontane .1 g
    Tetratriacontane .1 g
    Triacontane .1 g
    Tricosane .1 g
    n-Paraffin Mix C5,C6,C7,C8, (varied conc.), analytical standard
    n-Paraffin Mix C7,C8,C9,C10, (varied conc.), analytical standard
    n-Paraffin Mix C10,C12,C14,C16, certified reference material, (varied conc.)
    n-Paraffin Mix C18,C20,C22,C24, certified reference material, 2 % (w/w) each component in octane
    n-Paraffin Mix C22,C24,C28,C32, certified reference material, 2 % (w/w) each component in octane
    n-Paraffin Mix C24,C28,C32,C36, certified reference material, 2 % (w/w) each component in octane
    2,2,4-Trimethylpentane 1 g
    Eicosane 1 g
    See All (40)

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

Flash Point(F)

0.5 °F - closed cup

Flash Point(C)

-17.5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service