Skip to Content
Merck
All Photos(1)

Key Documents

442619

Supelco

Indole

analytical standard

Synonym(s):

1H-Benzo[b]pyrrole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7N
CAS Number:
Molecular Weight:
117.15
Beilstein:
107693
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

253-254 °C (lit.)

mp

51-54 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-30°C

SMILES string

c1ccc2[nH]ccc2c1

InChI

1S/C8H7N/c1-2-4-8-7(3-1)5-6-9-8/h1-6,9H

InChI key

SIKJAQJRHWYJAI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Indole is classified under the volatile flavor compounds (VFCs). It is known to play significant role in various biological functions such as anti-inflammatory, anticonvulsant, cardiovascular and antibacterial activities.

Application

Indole may be used as an analytical reference standard for the determination of the analyte in teas, crab meat spoilages, animal wastes and human waste by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

249.8 °F - closed cup

Flash Point(C)

121 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Discrimination of teas with different degrees of fermentation by SPME-GC analysis of the characteristic volatile flavour compounds.
Wang LF, et al.
Food Chemistry, 109(1), 196-206 (2008)
Analysis of crab meat volatiles as possible spoilage indicators for blue crab (Callinectes sapidus) meat by gas chromatography-mass spectrometry.
Sarnoski PJ, et al.
Food Chemistry, 122(3), 930-935 (2010)
Analysis of malodorous volatile substances of human waste: feces and urine.
Sato H, et al.
Journal of Health Science, 47(5), 483-490 (2001)
Experimental design to optimise the analysis of organic volatile compounds in cow slurry by headspace solid-phase microextraction-gas chromatography-mass spectrometry.
Larreta J, et al.
Journal of Chromatography A, 1136(1), 1-9 (2006)
Synthesis and antiinflammatory activity of heterocyclic indole derivatives.
Rani P, et al.
European Journal of Medicinal Chemistry, 39(5), 449-452 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service