Skip to Content
Merck
All Photos(1)

Key Documents

41900-U

Supelco

EPA TO-14 Calibration Mix 1 (39 components)

39 components, 100 ppb each component in nitrogen, pkg of 110 L (pi-marked cylinder), analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
77101502
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Agency

EPA TO-14

usage

sufficient for ≥80 uses

shelf life

1 yr

packaging

pkg of 110 L (pi-marked cylinder)

manufacturer/tradename

Scott Gas T9Z90012-110

concentration

100 ppb each component in nitrogen

parameter

1800 psi pressure

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

cylinder H × diam.

12 1/4 in. (31 cm) × 3 1/4 in. (8.3 cm)

fitting

CGA 180 fitting

application(s)

environmental

format

multi-component solution

Looking for similar products? Visit Product Comparison Guide

General description

Air Liquie America Specialty Gases LLC TO-14 gas blends are packaged in SCOTTY high pressure, transportable, laboratory-size cylinders. A SCOTTY cylinder contains over 100 liters of gas at 1800psi--enough gas to deliver adequate calibration for 80 or more uses. Each cylinder is equipped with CGA 180 fittings to plumb directly to your gas sampling system or through an appropriate regulator.

Where available, the components in Scott TO-14 gas calibration blends are traceable to NIST reference mixtures (NIST SRM 1804). All are certified for superior stability and accuracy.

The shelf life is one year.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

39 analytes in nitrogen

Analyte

Description

    Benzene
    Bromomethane
    Carbon tetrachloride
    Chlorobenzene
    Chloroethane
    Chloroform
    Chloromethane
    1,2-Dibromoethane
    1,2-Dichlorobenzene
    1,3-Dichlorobenzene
    1,4-Dichlorobenzene
    Dichlorodifluoromethane
    1,1-Dichloroethane
    1,2-Dichloroethane
    1,1-Dichloroethylene
    cis-1,2-Dichloroethylene
    Dichloromethane
    1,2-Dichloropropane
    cis-1,3-Dichloropropene
    trans-1,3-Dichloropropene
    1,2-Dichlorotetrafluoroethane
    Ethylbenzene
    Hexachloro-1,3-butadiene
    Styrene
    1,1,2,2-Tetrachloroethane
    Tetrachloroethylene
    Toluene
    1,2,4-Trichlorobenzene
    1,1,1-Trichloroethane
    1,1,2-Trichloroethane
    Trichloroethylene
    Trichlorofluoromethane
    1,1,2-Trichloro-1,2,2-trifluoroethane
    1,2,4-Trimethylbenzene
    1,3,5-Trimethylbenzene
    Vinyl chloride
    o-Xylene
    m-Xylene
    p-Xylene
    See All (39)

Pictograms

Gas cylinder

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Solvents are one of the three components of paints. They are petroleum-based chemicals which dissolve the pigment and binding agent for application.

Chromatograms

suitable for GC, application for air monitoring

Questions

  1. Is the concentration of 100 ppb for each component in the EPA TO-14 Calibration Mix 1 (39 components) based on weight/weight (w/w) or weight/volume (w/v)?

    1 answer
    1. It is weight by weight in moles.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service