Skip to Content
Merck
All Photos(1)

Key Documents

40079

Supelco

Phenanthrene solution

certified reference material, 5000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein:
1905428
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

SMILES string

c1ccc2c(c1)ccc3ccccc23

InChI

1S/C14H10/c1-3-7-13-11(5-1)9-10-12-6-2-4-8-14(12)13/h1-10H

InChI key

YNPNZTXNASCQKK-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Phenanthrene can be synthesized, via a multi-step synthesis process, using Haworth method involving, naphthalene and succinic anhydride as the starting materials. It can undergo chlorination in acetic acid to obtain a mixture of cis- and trans-9,10-dichloro-9,10-dihydrophenanthrenes as the major products.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hepworth.DJ, et al. et al.
Aromatic Chemistry, 13 (2002)
Jennifer R McKelvie et al.
Environmental pollution (Barking, Essex : 1987), 175, 75-81 (2013-01-23)
We evaluated the correlation between soil organic carbon (OC) content and metabolic responses of Eisenia fetida earthworms after exposure to phenanthrene (58 ± 3 mg/kg) spiked into seven artificial soils with OC contents ranging from 1 to 27% OC. Principal
Lei Wang et al.
Journal of hazardous materials, 244-245, 268-275 (2012-12-29)
To study the impact of organic matter (OM) properties on different sorption domain of hydrophobic organic contaminants, phenanthrene sorption on chemically modified geosorbents and synthesized charcoals (SCs) was investigated and explored using Dual-Mode model (DMM). Sorption of phenanthrene on two
Guohui Wang et al.
Environmental pollution (Barking, Essex : 1987), 175, 110-116 (2013-02-02)
Isosteric heats (ΔH) of sorption/desorption of phenanthrene were determined for carbonaceous materials (Pahokee peat, lignite, and high-volatile bituminous coal) and two soils based on reported equilibrium sorption/desorption isotherms at four different temperatures (4, 20, 46 and 77 °C). In addition, ΔH
Wenyan Li et al.
Environmental pollution (Barking, Essex : 1987), 176, 228-233 (2013-02-26)
Nanoparticles (NPs) in soils may participate in essential ecological services, since they have special characteristics arising from their nanoscale sizes and large surface areas. We did aqueous solubility enhancement experiments to derive the partition coefficients of phenanthrene between water and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service