Skip to Content
Merck
All Photos(1)

Key Documents

33111-U

Supelco

Vials, crimp top, serum bottle

volume 100 mL, clear glass bottle, O.D. × H 51.7 mm ( ) × 94.5 mm, pkg of 144 ea

Synonym(s):

Serum Bottle Crimp Vials

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001
NACRES:
NB.11
Pricing and availability is not currently available.

material

clear glass bottle

feature

autoclavable
closure type crimp top vial

packaging

pkg of 144 ea

O.D. × H

51.7 mm ( ) × 94.5 mm

volume

100 mL

fitting

thread for 20 mm

Looking for similar products? Visit Product Comparison Guide

General description

These vials are constructed of clear, borosilicate glass; closure not included.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service