27147
Vials, screw top with phenolic open-top cap, pre-assembled
volume 7 mL, clear glass vial, O.D. × H 17 mm × 60 mm, PTFE/red rubber septum
All Photos(1)
About This Item
UNSPSC Code:
41122001
Recommended Products
material
PTFE/red rubber septum
black phenolic cap (open-top)
clear glass vial
feature
closure type black phenolic hole cap
closure type screw top vial
packaging
pkg of 100 ea
O.D. × H
17 mm × 60 mm
volume
7 mL
fitting
thread for 15-425
Application
It may be used in headspace solid-phase microextraction (SPME) followed by GC-FID for simultaneous analysis of furan, 2-methylfuran and 2-pentylfuran from juice samples.
related product
replacement part
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
An efficient method for the simultaneous determination of furan, 2-methylfuran and 2-pentylfuran in fruit juices by headspace solid phase microextraction and gas chromatography?flame ionisation detector.
Hu, Gaofei, et al.
Food Chemistry, 192, 9-14 (2016)