23926
OMI® Tube Holder
for use with OMI-4 purifier tubes
Synonym(s):
OMI®-4 TUBE HOLDER
About This Item
Recommended Products
compatibility
for use with OMI-4 purifier tubes
Looking for similar products? Visit Product Comparison Guide
General description
- A re-useable tube holder that is installed into the gas delivery system. The use of polycarbonate provides see-through capability along with safety.
- A purifier tube containing an indicating resin (changes color from black to brown when exposed to as little as 1 ppm of moisture or oxygen). The use of glass provides see-through capability and prevents diffusion of room contaminants into the gas stream.
Spent purifier tubes are easily replaced. Simply unscrew the end assembly from the tube holder and replace it with a new purifier tube. The design prevents room air from entering the new tube during installation (protective foil covers on each end are only pierced as the end assembly is screwed back onto the tube holder to complete installation).
Note: This item requires a purifier tube (purchased separately).
Other Notes
- The OMI-2 tube holder is 10 in. (25.4 cm) long with a diameter of 1.5 in. (3.8 cm) and 1/8 in. fittings.
- The OMI-4 tube holder is 16 in. (40.6 cm) long with a diameter of 1.5 in. (3.8 cm) and 1/8 in. fittings.
Legal Information
required but not provided
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service