22269-U
Valco® Precision Sampling Syringe, Series A-2, Removable needle
volume 50 μL, bevel tip stainless steel needle (point style 2), needle L × O.D. × I.D. 2 in. × 0.028 in. × 0.005 in.
Synonym(s):
SYRINGE,PS-A-2,50UL
About This Item
Recommended Products
material
bevel tip stainless steel needle (point style 2)
description
Series A-2
feature
removable needle Series A2
packaging
pkg of 1 ea
manufacturer/tradename
VICI 050024
needle L × O.D. × I.D.
2 in. × 0.028 in. × 0.005 in.
volume
50 μL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Push-button valve - allows instantaneous injection
- Smaller volumes - great for small volatile samples
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Learn more about Biodiesel as a renewable, alternative diesel fuel produced from vegetable oils, animal fats, or recycled restaurant grease.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service