21742-U
Hamilton® needles for LT and TLL syringes
KEL-F hub, needle size 22 ga, needle L 51 mm (2 in.), pkg of 6 ea
About This Item
material
cone tip, side port stainless steel needle (point style 5)
needle size
22 ga
feature
KEL-F hub
packaging
pkg of 6 ea
manufacturer/tradename
Hamilton 90422
needle L
51 mm (2 in.)
compatibility
for use with Hamilton LT and TLL
Looking for similar products? Visit Product Comparison Guide
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service