Documents
20511
Supelco
Inlet Liner, for Packed GC (unpacked)
pkg of 25 ea
About This Item
UNSPSC Code:
41115719
packaging
pkg of 25 ea
technique(s)
gas chromatography (GC): suitable
L × O.D. × I.D.
91.5 mm × 3.0 mm × 1.8 mm
General description
The liner is the second most important part of the inlet. It is the centrepiece of the inlet system, basically a glass tube between the septum and the column, where the sample is evaporated and brought into the gas phase. There are three most important parameters one should keep in mind while choosing a liner, firstly liner volume, considering a typical liner has a volume of 900μl, secondly liner treatment, one should always use a deactivated liner, so that the glass does not react with the analytes introduced in the liner, thirdly liner design, different designs are available like straight, tapered, baffled and without glass wool and one should be careful enough while choosing the design such that it is compatible with the desired injection mode.
Deactivated (silanized) glass inlet liner for packed injection ports.
  • Fits Agilent (5890, 6890, and 7890)
Application
Inlet liner maybe used for the determination and evaluation of volatile components in food products. The vegetable oil maybe directly placed into the GC inlet liner packed with glass wool and then inserted into the heated inlet of the GC for analysis.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
Visit the Document Library
Navneet Sharma et al.
Life sciences, 168, 65-76 (2016-11-20)
p-tertbutylcalix[4]arene loaded nanoemulsion has been designed, characterized and evaluated for skin decontamination of radionuclides of interest in nuclear and radiological emergencies. Further, nanoemulsion was evaluated for Ex-vivo complexation, skin permeation, interaction and cytodermal toxicity. Ex-vivo skin complexation studies were conducted
Analysis of vegetable oils for flavor quality by direct gas chromatography.
Dupuy HP
Journal of the American Oil Chemists' Society, 54 (10), 445-449 (1977)
Fire Debris Analysis.
Stauffer E
Science, 249-253 (2007)
Julie Carnesecchi et al.
PloS one, 10(3), e0120672-e0120672 (2015-03-18)
The post-menopausal decrease in estrogen circulating levels results in rapid skin deterioration pointing out to a protective effect exerted by these hormones. The identity of the skin cell type responding to estrogens is unclear as are the cellular and molecular