20036
ORBO™ 77 H2SO4-Treated Carbon Bead (20/30), 500/250 mg
W,W,W separators, O.D. × L 8 mm × 110 mm, pkg of 50 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
W,W,W separators
carbon beads
Agency
OSHA ID-188
product line
ORBO™
composition
Bed A, 500 mg
Bed B, 250 mg
packaging
pkg of 50 ea
manufacturer/tradename
ORBO™ 77
technique(s)
active air sampling: suitable
O.D. × L
8 mm × 110 mm
matrix
H2SO4-treated carbon bead
particle size
20-30 mesh
application(s)
air monitoring
environmental
industrial hygiene
Related Categories
General description
- ORBO™ 77 H2SO4-Treated Carbon Bead (20/30), 500/250 mg solvent desorption tube is designed for gas and vapor sampling.
- It contains two beds of the same selective adsorbent separated by glass wool or foam.
- Dual-layer or two-bed construction of the tube allows for any sample breakthrough to be captured in the smaller back-up bed.
Legal Information
ORBO is a trademark of Sigma-Aldrich Co. LLC
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service