19144-U
Hamilton® needles for RN syringes, point style #5
needle size 26s ga, needle L 51 mm (2 in.), volume 2.5-100 μL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
cone tip, side port stainless steel needle
needle size
26s ga
feature
removable needle cone tip (side port)
packaging
pkg of 6 ea
manufacturer/tradename
Hamilton 7784-07
needle L
51 mm (2 in.)
volume
2.5-100 μL
Looking for similar products? Visit Product Comparison Guide
General description
Hamilton® needles for RN syringes, point style #5 are cone-tip needles equipped with side-port holes. Liquid samples can be filled and dispensed through the side hole, and septum damage is minimized by the solid domed tip.
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service