13026-U
Carbon Adsorbent Sampler Kit
matrix 60/80 GCB Kit
About This Item
Recommended Products
form
pellets
powder
packaging
kit of 6 bottles
technique(s)
LPLC: suitable
matrix
60/80 GCB Kit
Graphitized Carbon Black (GCB) adsorbents
matrix active group
carbon
particle size
60-80 mesh
application(s)
sample preparation
separation technique
reversed phase
Looking for similar products? Visit Product Comparison Guide
General description
- Must choose one or more to retain specific analyte(s)
- May be equally important to release analyte(s) during desorption
This convenient kit allows a cost-effective way to evaluate several of our specialty carbon adsorbents. Once you find an adsorbent or adsorbents that serve your purpose, we will work with you to either make your device for you, or to supply you bulk adsorbent (in a bottle, jar, or ampule; not packed inside a column or device).
For more information about any of our specialty carbon adsorbents, please visit sigma-aldrich.com/carbon
Other Notes
Footnote
Kit Components Only
- 60/80 Carbopack™ B 5 g
- 60/80 Carbopack C 5 g
- 60/80 Carbopack F 5 g
- 60/80 Carbopack X 5 g
- 60/80 Carbopack Y 5 g
- 60/80 Carbopack Z 5 g
Storage Class Code
11 - Combustible Solids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Synthetic CMS carbons offer tailored adsorbents for specific applications.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service