Skip to Content
Merck
All Photos(1)

Key Documents

01874

Supelco

Fenthion

reference material

Synonym(s):

O,O′-Dimethyl O″-[3-methyl-4-(methylthio)phenyl] thiophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15O3PS2
CAS Number:
Molecular Weight:
278.33
Beilstein:
1974129
MDL number:
UNSPSC Code:
41116107

grade

reference material

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

format

neat

storage temp.

2-8°C

SMILES string

COP(=S)(OC)Oc1ccc(SC)c(C)c1

InChI

1S/C10H15O3PS2/c1-8-7-9(5-6-10(8)16-4)13-14(15,11-2)12-3/h5-7H,1-4H3

InChI key

PNVJTZOFSHSLTO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This reference material (RM) is produced and certified in accordance with ISO/IEC 17025. This RM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Muta. 2 - STOT RE 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service